Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fumaric acid-13C4 | 201595-62-2 | 98.0% | 120.04 | 13C4H4O4 | 5 MG
SDP

Catalog No. 5000302113
Encompass_Preferred

Fumaric acid-13C4 is the 13C-labeled form of fumaric acid supplied as a white to off-white solid for use as a stable-isotope reference standard in metabolic and biochemical research. It is used in tracer experiments, metabolic profiling, and studies related to fumarase deficiency and oncometabolite pathways.

  • 13C-labeled stable-isotope reference standard for metabolic studies.
  • Stated chemical purity 98.0%.
  • Molecular formula 13C4H4O4; molecular weight 120.04.
  • Suitable for tracer experiments and metabolic profiling.
  • Available in small research pack sizes for analytical use.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.

Catalog No. 50-003-02113 Supplier Medchemexpress LLC Supplier No. HYW015883S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fumaric acid-13C4 is the 13C-labeled form of fumaric acid supplied as a white to off-white solid for use as a stable-isotope reference standard in metabolic and biochemical research. It is used in tracer experiments, metabolic profiling, and studies related to fumarase deficiency and oncometabolite pathways.

  • 13C-labeled stable-isotope reference standard for metabolic studies.
  • Stated chemical purity 98.0%.
  • Molecular formula 13C4H4O4; molecular weight 120.04.
  • Suitable for tracer experiments and metabolic profiling.
  • Available in small research pack sizes for analytical use.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.