Learn More
Medchemexpress LLC Fumaric acid-13C4 | 201595-62-2 | 98.0% | 120.04 g/mol | 13C4H4O4 | 10 MG
SDP
Catalog No. 5000303114
Encompass_Preferred
Fumaric acid-13C4 is the 13C-labeled stable-isotope form of fumaric acid used as an analytical standard and metabolic tracer in biochemical and pharmacological research. Supplied as a solid, high-purity labeled compound for in vitro and in vivo studies; intended for research use only.
  • High purity (98.0%) suitable for analytical work.
  • Isotopically labeled (13C4) for tracing and mass spectrometry applications.
  • Provided as a solid; standard pack size 10 mg.
  • Soluble in DMSO (50 mg/mL; may require ultrasonic) for stock solutions.
  • Compatible with common in vivo vehicle preparations at ≥2.5 mg/mL.
  • Useful as a reference standard for metabolic and flux analyses.
Catalog No. 50-003-03114 Supplier Medchemexpress LLC Supplier No. HYW015883S10MG
Fumaric acid-13C4 is the 13C-labeled stable-isotope form of fumaric acid used as an analytical standard and metabolic tracer in biochemical and pharmacological research. Supplied as a solid, high-purity labeled compound for in vitro and in vivo studies; intended for research use only.
  • High purity (98.0%) suitable for analytical work.
  • Isotopically labeled (13C4) for tracing and mass spectrometry applications.
  • Provided as a solid; standard pack size 10 mg.
  • Soluble in DMSO (50 mg/mL; may require ultrasonic) for stock solutions.
  • Compatible with common in vivo vehicle preparations at ≥2.5 mg/mL.
  • Useful as a reference standard for metabolic and flux analyses.