Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fumaric acid-13C4 | 201595-62-2 | MFCD00144377 | 98.0% | 120.04 | 13C4H4O4 | 1 MG
SDP

Catalog No. 5000300981
Encompass_Preferred

Fumaric acid-13C4 is the 13C-labeled form of fumaric acid supplied for laboratory research. It is intended for use in stable-isotope metabolic tracing, mass spectrometry, and metabolic flux analysis, and is provided with supporting documentation (datasheet, SDS, COA).

  • 98.0% purity.
  • Molecular formula 13C4H4O4 and molecular weight 120.04.
  • Suitable for stable-isotope metabolic tracing and mass spectrometry.
  • Provided with datasheet, safety data sheet, and certificate of analysis.
  • Available in small, pre-weighed quantities for tracer studies.

Catalog No. 50-003-00981 Supplier Medchemexpress LLC Supplier No. HYW015883S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fumaric acid-13C4 is the 13C-labeled form of fumaric acid supplied for laboratory research. It is intended for use in stable-isotope metabolic tracing, mass spectrometry, and metabolic flux analysis, and is provided with supporting documentation (datasheet, SDS, COA).

  • 98.0% purity.
  • Molecular formula 13C4H4O4 and molecular weight 120.04.
  • Suitable for stable-isotope metabolic tracing and mass spectrometry.
  • Provided with datasheet, safety data sheet, and certificate of analysis.
  • Available in small, pre-weighed quantities for tracer studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.