Learn More
Medchemexpress LLC Fumaric acid-d2 | 24461-32-3 | MFCD00144378 | 99.0% | 118.08 | C4H2D2O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW015883S3100MG
Encompass_Preferred
Fumaric acid-d2 is the deuterium-labeled analogue of fumaric acid (CAS 24461-32-3), intended for use as an isotopic standard in metabolic flux analysis, mass spectrometry quantitation, and tracer experiments. It is provided as a high-purity solid with documented solubility and storage recommendations for both powder and solution forms.
  • Deuterium-labeled isotopic standard for tracer studies.
  • High purity (99.0%).
  • Available in multiple pack sizes including 100 mg.
  • Soluble in DMSO ≥ 100 mg/mL for in vitro applications.
  • Documented storage stability for powder and prepared solutions.
Catalog No. 50-003-02115