Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fumaric acid-d2 | 24461-32-3 | MFCD00144378 | 99.0% | 118.08 g·mol⁻1 | C4H2D2O4 | 5 MG
SDP

Catalog No. 5000303627
Encompass_Preferred

Fumaric acid-d2 is the deuterium-labeled isotopologue of fumaric acid provided for laboratory research. It is used as a tracer or analytical standard in mechanistic and metabolic studies where deuterium labeling is required. The material is offered in small milligram quantities with high chemical and isotopic purity and is intended for research use only.

  • High isotopic purity (99.0%).
  • Chemical formula C4H2D2O4; molecular weight 118.08 g·mol⁻1.
  • High solubility in DMSO (≥100 mg/mL).
  • Available in small pack sizes suitable for analytical work.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years).
  • For research use only; not for human or clinical use.

Catalog No. 50-003-03627 Supplier Medchemexpress LLC Supplier No. HYW015883S35MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fumaric acid-d2 is the deuterium-labeled isotopologue of fumaric acid provided for laboratory research. It is used as a tracer or analytical standard in mechanistic and metabolic studies where deuterium labeling is required. The material is offered in small milligram quantities with high chemical and isotopic purity and is intended for research use only.

  • High isotopic purity (99.0%).
  • Chemical formula C4H2D2O4; molecular weight 118.08 g·mol⁻1.
  • High solubility in DMSO (≥100 mg/mL).
  • Available in small pack sizes suitable for analytical work.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years).
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.