Learn More
Medchemexpress LLC Fumaric acid-d2 | 24461-32-3 | MFCD00144378 | 99.0% | 118.08 g/mol | C4H2D2O4 | 25 MG
SDP
Catalog No. 5000303717
Encompass_Preferred
Fumaric acid-d2 is the deuterium-labeled analog of fumaric acid used as a stable isotope standard and research reagent. It is supplied as a white to off-white solid with 99.0% purity and is intended for analytical, metabolic, and tracer studies. For research use only.
  • Deuterium-labeled compound for tracer and analytical applications.
  • High purity (99.0%) suitable for reference standards.
  • Soluble in DMSO (≥ 100 mg/mL) for convenient stock preparation.
  • Stable when stored as a powder at -20°C for long-term storage.
  • Available in small mass sizes for analytical workflows.
Catalog No. 50-003-03717 Supplier Medchemexpress LLC Supplier No. 25MGHYW015883S3
Fumaric acid-d2 is the deuterium-labeled analog of fumaric acid used as a stable isotope standard and research reagent. It is supplied as a white to off-white solid with 99.0% purity and is intended for analytical, metabolic, and tracer studies. For research use only.
  • Deuterium-labeled compound for tracer and analytical applications.
  • High purity (99.0%) suitable for reference standards.
  • Soluble in DMSO (≥ 100 mg/mL) for convenient stock preparation.
  • Stable when stored as a powder at -20°C for long-term storage.
  • Available in small mass sizes for analytical workflows.