Learn More
Medchemexpress LLC Fumaric acid-d2 | 24461-32-3 | MFCD00144378 | 99.0% | 118.08 g·mol⁻¹ | C4H2D2O4 | 50 MG
SDP
Catalog No. 5000302869
Encompass_Preferred
Fumaric acid-d2 is the deuterium-labeled form of fumaric acid intended for use as a tracer and internal standard in quantitative analytical methods. It is suitable for NMR, GC-MS, and LC-MS applications and is used in research on fumarase deficiency and oncometabolites. The compound is supplied as a high-purity solid with documented solubility and storage recommendations.
  • Deuterium-labeled analog for tracer and internal standard use.
  • High listed purity suitable for analytical applications.
  • Soluble in DMSO; in vivo vehicle formulations are documented.
  • Stable solid with defined storage conditions for powder and solutions.
  • Available in multiple small packaging sizes for laboratory research.
Catalog No. 50-003-02869 Supplier Medchemexpress LLC Supplier No. HYW015883S350MG
Add to Cart
Add to Cart
Fumaric acid-d2 is the deuterium-labeled form of fumaric acid intended for use as a tracer and internal standard in quantitative analytical methods. It is suitable for NMR, GC-MS, and LC-MS applications and is used in research on fumarase deficiency and oncometabolites. The compound is supplied as a high-purity solid with documented solubility and storage recommendations.
  • Deuterium-labeled analog for tracer and internal standard use.
  • High listed purity suitable for analytical applications.
  • Soluble in DMSO; in vivo vehicle formulations are documented.
  • Stable solid with defined storage conditions for powder and solutions.
  • Available in multiple small packaging sizes for laboratory research.