Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fumaric acid-d4 | 194160-45-7 | 98.0% | 120.10 g·mol⁻1 | C4D4O4 | 25 MG
SDP

Catalog No. 5000302235
Encompass_Preferred

Fumaric acid-d4 is a deuterium-labeled analogue of fumaric acid supplied as a solid research reagent and isotopic standard for analytical and metabolic tracer studies. It provides reliable mass-shift labeling for LC-MS, NMR, and metabolic flux experiments.

  • Deuterium labeled (d4) fumaric acid for stable-isotope experiments.
  • High isotopic enrichment (~99.1%) enabling clear mass shifts.
  • HPLC purity 98% suitable for analytical applications.
  • Available in small milligram packages including 25 mg for low-quantity studies.
  • Molecular formula C4D4O4; molecular weight 120.10 g·mol⁻1.
  • Solid, white to off-white appearance; store powder at -20°C for long-term stability.

Catalog No. 50-003-02235 Supplier Medchemexpress LLC Supplier No. 25MGHYW015883S4
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fumaric acid-d4 is a deuterium-labeled analogue of fumaric acid supplied as a solid research reagent and isotopic standard for analytical and metabolic tracer studies. It provides reliable mass-shift labeling for LC-MS, NMR, and metabolic flux experiments.

  • Deuterium labeled (d4) fumaric acid for stable-isotope experiments.
  • High isotopic enrichment (~99.1%) enabling clear mass shifts.
  • HPLC purity 98% suitable for analytical applications.
  • Available in small milligram packages including 25 mg for low-quantity studies.
  • Molecular formula C4D4O4; molecular weight 120.10 g·mol⁻1.
  • Solid, white to off-white appearance; store powder at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.