Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fumaric acid-d4 | 194160-45-7 | MFCD00190450 | 98.0% | 120.10 g·mol^-1 | C4D4O4 | 50 MG
SDP

Catalog No. 5000299623
Encompass_Preferred

Fumaric acid-d4 is the deuterium-labeled analogue of fumaric acid supplied as a stable isotope standard for analytical and research use. It is intended for metabolomics, tracer studies, and mass spectrometry assays requiring isotopically labeled internal standards, and supports investigations into fumarase deficiency and oncometabolite pathways.

  • Deuterium-labeled internal standard for analytical assays.
  • Molecular formula C4D4O4, molecular weight 120.10 g·mol^-1.
  • High isotopic enrichment suitable for mass spectrometry.
  • Available in small pack sizes appropriate for research workflows.
  • Used in metabolomics, tracer, and biochemical pathway studies.

Catalog No. 50-002-99623 Supplier Medchemexpress LLC Supplier No. HYW015883S450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fumaric acid-d4 is the deuterium-labeled analogue of fumaric acid supplied as a stable isotope standard for analytical and research use. It is intended for metabolomics, tracer studies, and mass spectrometry assays requiring isotopically labeled internal standards, and supports investigations into fumarase deficiency and oncometabolite pathways.

  • Deuterium-labeled internal standard for analytical assays.
  • Molecular formula C4D4O4, molecular weight 120.10 g·mol^-1.
  • High isotopic enrichment suitable for mass spectrometry.
  • Available in small pack sizes appropriate for research workflows.
  • Used in metabolomics, tracer, and biochemical pathway studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.