Learn More
HIMEDIA Fumaric Acid, Hi-AR, GRM1231, Laboratory Chemicals, Organic Acid, Analytical Reagent, CAS 110-17-8, Acidulant and Reagent, Powder, 100 g
Catalog No. NC4013640
Encompass_Preferred
Fumaric acid, Hi-AR is an analytical reagent grade dicarboxylic acid (C4H4O4, CAS 110-17-8, molecular weight 116.07) supplied as white to off-white crystals, powder or lumps whose FTIR matches the standard reference pattern. Soluble in ethanol, it serves as an analytical reagent, an acidulant and antioxidant, and a versatile intermediate and standard in laboratory and synthetic chemistry. Store below 30 C, shelf life four years. Packaged as 100 g.
Catalog No. NC4013640 Supplier HIMEDIA Supplier No. GRM1231100G
Fumaric acid, Hi-AR is an analytical reagent grade dicarboxylic acid (C4H4O4, CAS 110-17-8, molecular weight 116.07) supplied as white to off-white crystals, powder or lumps whose FTIR matches the standard reference pattern. Soluble in ethanol, it serves as an analytical reagent, an acidulant and antioxidant, and a versatile intermediate and standard in laboratory and synthetic chemistry. Store below 30 C, shelf life four years. Packaged as 100 g.