Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fumaric acid monoethyl ester, 95%

Catalog No. AC156200250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156200250 25 g Glass bottle
AC156201000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC156200250 Supplier Thermo Scientific Chemicals Supplier No. 156200250
Add to Cart

CAS: 2459-05-4 | C6H8O4 | 144.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5-4-2459
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.13
MDL Number MFCD00002699
InChI Key XLYMOEINVGRTEX-ONEGZZNKSA-N
Synonym Monoethylefumarate
IUPAC Name (2E)-4-ethoxy-4-oxobut-2-enoic acid
SMILES CCOC(=O)\C=C\C(O)=O

Specifications

Melting Point 64°C to 69°C
Color Beige to White
Boiling Point 147°C (16.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula HO2CCH=CHCO2C2H5
Quantity 25 g
Beilstein 02, 741
Formula Weight 144.13
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Fumaric acid monoethyl ester
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 219-544-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.