Learn More
U.S. Pharmacopeia Fumaric Acid, 110-17-8, MFCD00002700, 200mg
Catalog No. NC0419333
Encompass
Molecular formula C4H4O4, Molecular weight 116.07, Melting Point 546.8 - 548.6 °F (286 - 287 °C) (sublimes at 200 ° C ), Boiling Point 554 °F (290 °C),
Catalog No. NC0419333 Supplier U.S. Pharmacopeia Supplier No. 1286708
Molecular formula C4H4O4, Molecular weight 116.07, Melting Point 546.8 - 548.6 °F (286 - 287 °C) (sublimes at 200 ° C ), Boiling Point 554 °F (290 °C),