Learn More
Air Science Usa Fume Adsorber, Configurable as Horizontal or Vertical Flow Versions, 115V 60Hz
SDP
Catalog No. NC1125736
Encompass_Preferred
Fume Box (AP60) ductless enclosures are designed to protect the user from chemicals, vapors or powders during low-volume chemical manipulations by effectively containing low concentrations of noxious fumes, vapors or powders. Requires qty. 1 AP60-Xxx Main Filter sold separately.Ductless Technology Advanced carbon filtration technology offers a safe, high performance alternative to conventional ducted fume hoods for a broad range of applications
Catalog No. NC1125736 Supplier Air Science Usa Supplier No. AP60H
Fume Box (AP60) ductless enclosures are designed to protect the user from chemicals, vapors or powders during low-volume chemical manipulations by effectively containing low concentrations of noxious fumes, vapors or powders. Requires qty. 1 AP60-Xxx Main Filter sold separately.Ductless Technology Advanced carbon filtration technology offers a safe, high performance alternative to conventional ducted fume hoods for a broad range of applications