Learn More
Air Science Usa Fume Adsorber XL, Horizontal Flow, 115V 60Hz
SDP
Catalog No. NC1837396
Encompass_Preferred
Fume Box (AP60-XL) ductless enclosures are designed to protect the user from chemicals, vapors or powders during low-volume chemical manipulations by effectively containing low concentrations of noxious fumes, vapors or powders. Requires qty. 1 AP60-Xxx Main Filter sold separately
Catalog No. NC1837396 Supplier Air Science Usa Supplier No. AP60XL
Fume Box (AP60-XL) ductless enclosures are designed to protect the user from chemicals, vapors or powders during low-volume chemical manipulations by effectively containing low concentrations of noxious fumes, vapors or powders. Requires qty. 1 AP60-Xxx Main Filter sold separately