Learn More
Mopec Fume Capture Support Frame
SDP
Catalog No. 22000154 Shop All Mopec Products
Change view
Click to view available options
For Use With (Equipment):
BF710
Catalog No. For Use With (Equipment)
22-000-154 BF710
1 options
Catalog No. 22-000-154 Supplier Mopec Supplier No. BF736
Add to Cart
Add to Cart
Fume hoods are an excellent solution for extracting fumes from the encapsulated work area. Strong, lightweight and portable, Mopec’s manufactured hoods are designed to isolate and remove irritating, potentially hazardous organic vapors from the laboratory environment.
Construction:
  • For use with BF710 Self-Contained Exhaust ModuleBF720 Ventilation Adapter
Dimensions:
  • 36 in. L x 20 in. H x 20 in. W
Material:
  • Extruded aluminum frame construction and clear high impact lexan™
Specifications
For Use With (Equipment) BF710
Product Type Support Frame
For Use With (Application) Fume Capture Support Frame