Description
- Includes (1) fume extractor
- Made to go on top of any countertop or work surface
- Aluminum frame, clear polycarbonate, ABS plastic, stainless steel screws
- Requires a Qty of 2 of either BF720 (connects to facility ventilation) or BF710 (recirculating/self contained blower) to function
- Options and Accessories
- BF724, BF736, fume extraction modules
- BF710 Fume extraction module
- BF720 Fume extractor duct adapter
- PM0603 vent duct (sold separately by the foot)
- PB0378 clamp for vent duct (sold separately, individually)
- BF002, BF003 (filters for BF710, Qty of 1 Ea. required as replacements)
- Connect your fume extractor to the building ventilation system to remove fumes
- Place a BF710 fume exhaust module on top to filter air to reciruculate back into the room
Specifications
Specifications
| Product Type | Benchtop Fume Hood |
| Coating | Uncoated |
| Filter Type | Unfiltered |
| Lighting Type | None |
| Material | Aluminum, Polycarbonate, Stainless Steel |
| Dimensions (L x W x H) | 48 x 20 x 20 in. |
| IP Rating | None |