Learn More
Mopec Fume Extractor
SDP
Catalog No. 22445899 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W x H):
48 x 20 x 20 in.
60 x 20 x 20 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
22-445-899 60 x 20 x 20 in.
22-445-898 48 x 20 x 20 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-445-899 Supplier Mopec Supplier No. BF760
Add to Cart
Add to Cart
Casework Line features durable, modular stainless-steel cabinetry engineered to meet the distinct needs for clinical workspaces
  • Includes (1) fume extractor
  • Made to go on top of any countertop or work surface
  • Aluminum frame, clear polycarbonate, ABS plastic, stainless steel screws
  • Requires a Qty of 2 of either BF720 (connects to facility ventilation) or BF710 (recirculating/self contained blower) to function
  • Options and Accessories
    • BF724, BF736, fume extraction modules
    • BF710 Fume extraction module
    • BF720 Fume extractor duct adapter
    • PM0603 vent duct (sold separately by the foot)
    • PB0378 clamp for vent duct (sold separately, individually)
    • BF002, BF003 (filters for BF710, Qty of 1 Ea. required as replacements)
  • Connect your fume extractor to the building ventilation system to remove fumes
  • Place a BF710 fume exhaust module on top to filter air to reciruculate back into the room
Specifications
Product Type Benchtop Fume Hood
Coating Uncoated
Filter Type Unfiltered
Lighting Type None
Material Aluminum, Polycarbonate, Stainless Steel
Dimensions (L x W x H) 60 x 20 x 20 in.
IP Rating None