Learn More
Air Science Usa Fume Hood 49" Wide Ducted High Clearance Laboratory Exhaust Hood for Safe Removal of Hazardous Fumes, Vapors, and Particulates
SDP
Catalog No. 5000580918
Encompass_Preferred
Ducted Advanced Fume Hoods / High Clearance Enclosures Designed to safely exhaust harmful fumes and particulates away from the operator when connected to an external ventilation system. Purair Ducted Advanced High Clearance Fume Hoods provide enhanced internal height, width, and depth compared to standard models, allowing greater flexibility for larger equipment and demanding applications. Constructed with a chemical-resistant epoxy-coated steel frame, these units offer durability and long-term performance. Clear panels on all sides deliver 360° visibility and allow ambient light into the workspace, while the hinged front sash provides easy access and containment. Installed as a fixed system, these hoods create negative pressure to effectively capture and exhaust hazardous vapors to the outside. Adjustable face velocity allows control over airflow balance and energy efficiency
Catalog No. 50-005-80918 Supplier Air Science Usa Supplier No. P20XTENCLDUCT
Ducted Advanced Fume Hoods / High Clearance Enclosures Designed to safely exhaust harmful fumes and particulates away from the operator when connected to an external ventilation system. Purair Ducted Advanced High Clearance Fume Hoods provide enhanced internal height, width, and depth compared to standard models, allowing greater flexibility for larger equipment and demanding applications. Constructed with a chemical-resistant epoxy-coated steel frame, these units offer durability and long-term performance. Clear panels on all sides deliver 360° visibility and allow ambient light into the workspace, while the hinged front sash provides easy access and containment. Installed as a fixed system, these hoods create negative pressure to effectively capture and exhaust hazardous vapors to the outside. Adjustable face velocity allows control over airflow balance and energy efficiency