Learn More
Perkin Elmer US LLC Fume Hood II for Autosampler
Catalog No. 502103743
Encompass_Preferred
Made from clear, acid resistant polycarbonate with a solid surface material base, the fume hood eliminates the risk of trace-metal contamination common with traditional fume hoods. HEPA filters ensure clean input air. The fume hood can be connected to an in-house laboratory exhaust system or used with the SPB blower unit (N9308022). Fume hoods come flatpacked to save transport costs and can be assembled in minutes without tools.
Catalog No. 50-210-3743 Supplier Perkin Elmer US LLC Supplier No. N9308036
Add to Cart
Add to Cart
Request a Quote
Made from clear, acid resistant polycarbonate with a solid surface material base, the fume hood eliminates the risk of trace-metal contamination common with traditional fume hoods. HEPA filters ensure clean input air. The fume hood can be connected to an in-house laboratory exhaust system or used with the SPB blower unit (N9308022). Fume hoods come flatpacked to save transport costs and can be assembled in minutes without tools.