Learn More
Electron Microscopy Sciences Fume Hood Shelves, Medium
SDP
Catalog No. 5032747
Encompass_Preferred
Special designed fume hood shelves to offer addition storage space for your lab. They are designed to maintain air-flow and keep hazardous materials and other items off hood floor for safety.
Size: 48" L x 5" W
Height: 8"
Catalog No. 50-327-47 Supplier Electron Microscopy Sciences Supplier No. 6439602
Special designed fume hood shelves to offer addition storage space for your lab. They are designed to maintain air-flow and keep hazardous materials and other items off hood floor for safety.
Size: 48" L x 5" W
Height: 8"