Learn More
Medchemexpress LLC HY-N2143 1mg Medchemexpress, Fumitremorgin C CAS:118974-02-0 Purity:>98%
SDP
Catalog No. 501933889
Encompass_Preferred
Medchemexpress, HY-N2143 1mg Fumitremorgin C CAS:118974-02-0 Fumitremorgin C is a potent and selective ABCG2/BRCP inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3889 Supplier Medchemexpress LLC Supplier No. HYN21431MG
Medchemexpress, HY-N2143 1mg Fumitremorgin C CAS:118974-02-0 Fumitremorgin C is a potent and selective ABCG2/BRCP inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.