Learn More
AdipoGen Fumitremorgin C
SDP
Supplier:  AdipoGen BVT0189M001
Encompass_Preferred
Chemical. CAS 118974-02-0. Formula C22H25N3O3. MW 379.5. Isolated from Aspergillus fumigatus. Mycotoxin. Cytotoxic. Tremorgenic. Potent and specific inhibitor of the breast cancer resistance protein BCRP ABCG2. Reverses multidrug resistance mediated by BCRP and increases cytotoxicity of several anticancer agents in vitro.
Catalog No. 50-256-2075