Learn More
Abcam Fumitremorgin C (Tryptoquivaline), Chemosensitizing agent, 1MG
Catalog No. 502705941
Encompass_Preferred
MW 379.5 Da, Purity >98%. Chemosensitizing agent. Potent and selective BCRP inhibitor. Selective AKT inhibitor. Selectively reverses resistance to and potentiates mitoxantrone (ab145668), doxorubicin (ab120629) and topotecan (ab120388) in BCRP-transfected cells. Mycotoxin. Shows antiproliferative effects. Shows tremorgenic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5941 Supplier Abcam Supplier No. AB1442581MG
MW 379.5 Da, Purity >98%. Chemosensitizing agent. Potent and selective BCRP inhibitor. Selective AKT inhibitor. Selectively reverses resistance to and potentiates mitoxantrone (ab145668), doxorubicin (ab120629) and topotecan (ab120388) in BCRP-transfected cells. Mycotoxin. Shows antiproliferative effects. Shows tremorgenic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement