Learn More
Abcam Fumitremorgin C (Tryptoquivaline), Chemosensitizing agent, 250UG
Catalog No. 502705940
Encompass_Preferred
MW 379.5 Da, Purity >98%. Chemosensitizing agent. Potent and selective BCRP inhibitor. Selective AKT inhibitor. Selectively reverses resistance to and potentiates mitoxantrone (ab145668), doxorubicin (ab120629) and topotecan (ab120388) in BCRP-transfected cells. Mycotoxin. Shows antiproliferative effects. Shows tremorgenic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5940 Supplier Abcam Supplier No. AB144258250UG
MW 379.5 Da, Purity >98%. Chemosensitizing agent. Potent and selective BCRP inhibitor. Selective AKT inhibitor. Selectively reverses resistance to and potentiates mitoxantrone (ab145668), doxorubicin (ab120629) and topotecan (ab120388) in BCRP-transfected cells. Mycotoxin. Shows antiproliferative effects. Shows tremorgenic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement