Learn More
Cayman Chemical Fumonisin B1-13C34, Purity: ≥95%, CAS: 1217458-62-2, 1.2 mL
SDP
Catalog No. 502991514
Encompass_Preferred
An internal standard for the quantification of fumonisin B1 by GC- or LC-MS
Catalog No. 50-299-1514 Supplier Cayman Chemical Supplier No. 312721.2 ML
An internal standard for the quantification of fumonisin B1 by GC- or LC-MS