Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fumonisin B1-13C34 Solution, 25 μg/mL in Acetonitrile: water, MilliporeSigma™ Supelco™

Catalog No. 111017940 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7940 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7940 Supplier MilliporeSigma Supelco Supplier No. 336211ML
Add to Cart
Add to Cart

Certan Vial

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 1217458-62-2
Flash Point 2°C
Linear Formula 13C34H59 NO15
Type Analytical Standard
Quantity 1 mL
Formula Weight 755.58
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 25 μg/mL in acetonitrile: water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.