Learn More
Cayman Chemical Fumonisin B1, Purity: ≥95%, CAS: 116355-83-0, 1 mg
SDP
Catalog No. 502896958
Encompass_Preferred
A mycotoxin; inhibits ceramide synthase (IC50 = 0.1 µM for the rat liver enzyme); induces DNA fragmentation and apoptosis in CV-1 cells at 5 and 25 µM, respectively; cytotoxic to primary rat hepatocytes; induces hepatocyte nodule formation, a marker of cancer initiation, in rats at 500 mg/kg for 21 days; has been detected in corn and corn-based foods and livestock feeds
Catalog No. 50-289-6958 Supplier Cayman Chemical Supplier No. 625801 MG
A mycotoxin; inhibits ceramide synthase (IC50 = 0.1 µM for the rat liver enzyme); induces DNA fragmentation and apoptosis in CV-1 cells at 5 and 25 µM, respectively; cytotoxic to primary rat hepatocytes; induces hepatocyte nodule formation, a marker of cancer initiation, in rats at 500 mg/kg for 21 days; has been detected in corn and corn-based foods and livestock feeds