Learn More
Cusabio Technology LLC Fumonisin B1 ELISA Kit
Catalog No. 5000050111
Encompass_Preferred
This ELISA kit detects Fumonisin B1 in feed and corn samples with high specificity and sensitivity Designed for quantitative analysis it offers a reliable detection range of 3 ppb to 243 ppb making it suitable for monitoring contamination levels in agricultural products The kit features a low detection limit of 3 ppb ensuring accurate identification of trace amounts of Fumonisin B1 to meet food safety standards Its optimized protocol requires minimal sample preparation and delivers consistent results within standard assay timelines Ideal for laboratories and quality control departments this kit provides a cost-effective solution for routine screening of mycotoxin contamination in grain-based commodities
Catalog No. 50-000-50111 Supplier Cusabio Technology LLC Supplier No. CSBE11104
This ELISA kit detects Fumonisin B1 in feed and corn samples with high specificity and sensitivity Designed for quantitative analysis it offers a reliable detection range of 3 ppb to 243 ppb making it suitable for monitoring contamination levels in agricultural products The kit features a low detection limit of 3 ppb ensuring accurate identification of trace amounts of Fumonisin B1 to meet food safety standards Its optimized protocol requires minimal sample preparation and delivers consistent results within standard assay timelines Ideal for laboratories and quality control departments this kit provides a cost-effective solution for routine screening of mycotoxin contamination in grain-based commodities