Learn More
Cayman Chemical Fumonisin B2, Purity: ≥95%, CAS: 116355-84-1, 10 mg
SDP
Catalog No. 502952329
Encompass_Preferred
A mycotoxin; cytotoxic against a panel of seven rat hepatoma cell lines (IC50s = 3-50 µg/ml), as well as MDCK cells (IC50 = 2 µg/ml); induces hepatocyte nodule formation in rats at 1,000 mg/kg; has been detected in corn and corn-based foods and livestock feeds
Catalog No. 50-295-2329 Supplier Cayman Chemical Supplier No. 1322710 MG
A mycotoxin; cytotoxic against a panel of seven rat hepatoma cell lines (IC50s = 3-50 µg/ml), as well as MDCK cells (IC50 = 2 µg/ml); induces hepatocyte nodule formation in rats at 1,000 mg/kg; has been detected in corn and corn-based foods and livestock feeds