Learn More
Enzo Life Sciences Fumonisin B2 (1 mg). CAS: 116355-84-1
SDP
Supplier:  Enzo Life Sciences BMLSL2190001
Encompass_Preferred
Mycotoxin, structurally similar to fumonisin B1 (BML-SL220). Found as contaminant mostly in corn silage; was detected recently for the first time in the industrially important Aspergillus niger. Induces cancer. Inducer of apoptosis. Inhibits sphinganine N-acyltransferase (ceramide synthase) at IC50 = 0.1µM. Inhibits de novo sphingolipid biosynthesis. Purity: ≥98% (TLC). Solubility: Soluble in water (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2128