Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fumonisin B3 Solution, 50 μg/mL in Acetonitrile: water, MilliporeSigma™ Supelco™

Catalog No. 111017983 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7983 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7983 Supplier MilliporeSigma Supelco Supplier No. 326061ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certan Vial

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 136379-59-4
Flash Point 6°C - not determined
Linear Formula C34H59 NO14
MDL Number MFCD08458745
Solubility Soluble in water
Type Analytical Standard
Quantity 1 mL
Formula Weight 705.83
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 50 μg/mL in acetonitrile: water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.