Learn More
Sigma Aldrich Fine Chemicals Biosciences fumonisin Mixture 50 mug/
Catalog No. 502900192
Encompass
fumonisin mixture comprises of fumonisins B1 and B2 a group of nongenotoxic carcinogens produced primarily by the fungus fusarium moniliforme.
Catalog No. 50-290-0192 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 341431ML
fumonisin mixture comprises of fumonisins B1 and B2 a group of nongenotoxic carcinogens produced primarily by the fungus fusarium moniliforme.