Learn More
AdipoGen Funalenone (1 mg)
SDP
Supplier:  AdipoGen AGCN20137M001
Encompass_Preferred
Chemical. CAS: 259728-61-5. Formula: C15H12O6. MW: 288.3. Isolated from Aspergillus niger. Matrix metalloproteinase-1 (MMP-1; Type I collagenase) inhibitor. HIV-1 integrase inhibitor.
Catalog No. 50-205-3512