Learn More
Cayman Chemical β-Funaltrexamine (hydrochloride), Purity: ≥95%, CAS: 72786-10-8, 10 mg
SDP
Catalog No. 502942055
Encompass_Preferred
A μ- and κ1-opioid receptor antagonist (Kis = 0.3 and 0.2 nM, respectively); selective for μ- and κ1-opioid receptors over the δ-opioid receptor (Ki = 12.8 nM); reversibly inhibits electricity-induced twitches of isolated guinea pig ileal longitudinal muscle strips (IC50 = 48 nM), indicating opioid agonist activity; inhibits endomorphin 1-induced conditioned place preference in mice at 0.3 µg/animal, i.c.v.; prevents decreases in urine output induced by fentanyl, D-propoxyphene, buprenorphine, profadol, or bromadoline in water-loaded rats at 40 mg/kg; reduces hemokinin 1-induced increases in the latency to withdrawal in the tail-flick test in mice
Catalog No. 50-294-2055 Supplier Cayman Chemical Supplier No. 3990110 MG
A μ- and κ1-opioid receptor antagonist (Kis = 0.3 and 0.2 nM, respectively); selective for μ- and κ1-opioid receptors over the δ-opioid receptor (Ki = 12.8 nM); reversibly inhibits electricity-induced twitches of isolated guinea pig ileal longitudinal muscle strips (IC50 = 48 nM), indicating opioid agonist activity; inhibits endomorphin 1-induced conditioned place preference in mice at 0.3 µg/animal, i.c.v.; prevents decreases in urine output induced by fentanyl, D-propoxyphene, buprenorphine, profadol, or bromadoline in water-loaded rats at 40 mg/kg; reduces hemokinin 1-induced increases in the latency to withdrawal in the tail-flick test in mice