Learn More
Thermo Scientific Guardcap H Vial Cap | | Pack of 125
Supplier:  Thermo Scientific 302504
Encompass
Remove high levels of alkaline earth and transition metals, neutralize basic samples, and filter particulates. Designed for use with Thermo Scientific Dionex AS-DV Autosampler PolyVials. Contain sulfonated strong cation exchange resin in the hydronium form embedded in a high density polyethylene matrix. Seamlessly integrate automated inline sample preparation into existing sample workflow without additional labor, time, and complexity.
Catalog No. NC1353426