Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Fundamentals of Electronics Kit
GREENER_CHOICESCI_ED

Catalog No. S160141 Shop All Supertek Scientific Products
Change view
Click to view available options
Class Size:
1 to 2 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S160141 1 to 2 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S160141 Supplier Supertek™ Supplier No. PH92050SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Supertek™ Fundamentals of Electronics Kit is a comprehensive tool for introducing students to Ohm's Law, resistors, capacitors, semiconductors, and transistors. It even comes with a top-notch solderless breadboard!

  • Explains basic concepts of electronics.
  • High quality solderless breadboard.
  • Easy to build and learn.
  • Symbolic representation of components.
  • Color coding chart to read resistor values.

Specifications

Grade 9 to Undergraduate
Includes Breadboard on base, LEDs (Red and Green), Capacitors, Resistors, Switches, Buzzer, Transistors (NPN and PNP), Diode, Photoresistor, Connecting wires, Battery, Instruction manual
Class Size 1 to 2 Lab Groups
Product Type Electronics Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.