Learn More
Cayman Chemical Fungal Lysergol 100mg
SDP
Supplier:  Cayman Chemical 11717100 MG
Encompass_Preferred
An ergot alkaloid and 5-HT1A, 5-HT2A, and 5-HT2C receptor agonist (EC50s = 73, 1.6, and 6.6 nM, respectively, in a FLIPR assay); induces contractions in isolated rat tail arteries (EC50 = 436.5 nM); enhances the antibacterial activity of nalidixic acid against susceptible and drug-resistant E. coli at 10 UG/ml
Catalog No. 50-295-2311