Learn More
Cayman Chemical Fungal Lysergol 50mg
SDP
Catalog No. 502951765
Encompass_Preferred
An ergot alkaloid and 5-HT1A, 5-HT2A, and 5-HT2C receptor agonist (EC50s = 73, 1.6, and 6.6 nM, respectively, in a FLIPR assay); induces contractions in isolated rat tail arteries (EC50 = 436.5 nM); enhances the antibacterial activity of nalidixic acid against susceptible and drug-resistant E. coli at 10 UG/ml
Catalog No. 50-295-1765 Supplier Cayman Chemical Supplier No. 1171750 MG
An ergot alkaloid and 5-HT1A, 5-HT2A, and 5-HT2C receptor agonist (EC50s = 73, 1.6, and 6.6 nM, respectively, in a FLIPR assay); induces contractions in isolated rat tail arteries (EC50 = 436.5 nM); enhances the antibacterial activity of nalidixic acid against susceptible and drug-resistant E. coli at 10 UG/ml