Learn More
Cayman Chemical Asperfuran, Purity: ≥95%, CAS: 129277-10-7, 2.5 mg
SDP
Catalog No. 502989479
Encompass_Preferred
A fungal metabolite; inhibits the growth of 23 fungi in a disc assay at 50 µg per disc; inhibits chitin synthase (IC50 = 300 µM); inhibits proliferation of HeLa S3 and L1210 cancer cells (IC50s = 25 µg/ml for both)
Catalog No. 50-298-9479 Supplier Cayman Chemical Supplier No. 299892.5 MG
A fungal metabolite; inhibits the growth of 23 fungi in a disc assay at 50 µg per disc; inhibits chitin synthase (IC50 = 300 µM); inhibits proliferation of HeLa S3 and L1210 cancer cells (IC50s = 25 µg/ml for both)