Learn More
Polysciences, Inc. FUNGI FLUOR KIT - fluorescent stain/counterstain procedure for various fungal organisms. The kit can be used to screen a variety of specimen types from sputum to skin scrapings for fungal detection.
SDP
Catalog No. NC9020325
Encompass_Preferred
This kit offers a quick fluorescent stain/counterstain procedure for various fungal organisms. The kit can be used to screen a variety of specimen types from sputum to skin scrapings for fungal detection.
Catalog No. NC9020325 Supplier Polysciences, Inc. Supplier No. 174421
This kit offers a quick fluorescent stain/counterstain procedure for various fungal organisms. The kit can be used to screen a variety of specimen types from sputum to skin scrapings for fungal detection.