Learn More
Cayman Chemical Enniatin B, Purity: ≥95%, CAS: 917-13-5, 1 mg
SDP
Catalog No. 502969096
Encompass_Preferred
Inhibits the Pdr5p in yeast, augmenting, at concentrations as low as 0.8 μM, the ability of cerulenin or cycloheximide to impair cell proliferation; inhibits ACAT (IC50 = 113 μM), blocking cholesteryl ester formation
Catalog No. 50-296-9096 Supplier Cayman Chemical Supplier No. 153821 MG
Inhibits the Pdr5p in yeast, augmenting, at concentrations as low as 0.8 μM, the ability of cerulenin or cycloheximide to impair cell proliferation; inhibits ACAT (IC50 = 113 μM), blocking cholesteryl ester formation