Learn More
Cayman Chemical Fuscin, Purity: ≥95%, CAS: 83-85-2, 10 mg
SDP
Catalog No. 502941221
Encompass_Preferred
A fungal metabolite with diverse biological activities; inhibits binding of atractyloside to rat liver mitochondria in an ADP-dependent manner at 50 μM in a radioligand binding assay; reduces the glutathione content of rat liver mitochondria to 28% of controls at 20 μM; inhibits NADH oxidation in sonicated pigeon heart mitochondria preparations; competes with MIP-1α for binding to CCR5 chemokine receptors in vitro (IC50 = 21 μM)
Catalog No. 50-294-1221 Supplier Cayman Chemical Supplier No. 2748510 MG
A fungal metabolite with diverse biological activities; inhibits binding of atractyloside to rat liver mitochondria in an ADP-dependent manner at 50 μM in a radioligand binding assay; reduces the glutathione content of rat liver mitochondria to 28% of controls at 20 μM; inhibits NADH oxidation in sonicated pigeon heart mitochondria preparations; competes with MIP-1α for binding to CCR5 chemokine receptors in vitro (IC50 = 21 μM)