Learn More
Cayman Chemical Lachnone A, Purity: ≥95%, CAS: 903892-99-9, 2.5 mg
SDP
Catalog No. 502936907
Encompass_Preferred
A fungal metabolite; inactive against P. falciparum, C. albicans, and M. tuberculosis; not cytotoxic to KB or BC-1 cells
Catalog No. 50-293-6907 Supplier Cayman Chemical Supplier No. 264622.5 MG
A fungal metabolite; inactive against P. falciparum, C. albicans, and M. tuberculosis; not cytotoxic to KB or BC-1 cells