Learn More
StatLab Fungi-Tape Pre-Cut Cover Tape for Mycology Preparations
SDP
Catalog No. 22130486
Change view
Click to view available options
Product Type:
Fungi Tape
Catalog No. Product Type
22-130-486 Fungi Tape
1 options
This item is not returnable. View return policy
Catalog No. 22-130-486 Supplier StatLab Supplier No. 745
Add to Cart
Add to Cart
This item is not returnable. View return policy
Adhesive tape for quickly creating slide mounts of fungi from primary cultures
  • Pliable enough to make a loop and take a sample from your culture
  • Each roll comes in a dispensing box for easy use and contains enough tape for approximately 360 coverslips
  • Fungi-Tape can be used with MycoPerm Blue and a microscope slide to create a long lasting fungi mount
  • 1 roll each
Specifications
Quantity 1 Roll
Product Type Fungi Tape