Description
Fungi TISSUE-TROL™ Control Slides are meticulously prepared histology control slides featuring paraffin-embedded tissue sections from mouse lungs containing Candida albicans. These control slides are essential for pathology laboratories in providing support for quality assurance and accurate fungal identification in histological samples.
Specifications
Specifications
| Product Type | Control Slide |