Learn More
Bioworld Fungi/Yeast Genomic DNA Isolation Kit, 50 Prep(s)
Catalog No. 50196955
Encompass_Preferred
For the rapid purification of DNA from yeast cells and fungal spores or mycelium No phenol required for isolation Purified DNA fully compatible with sensitive downstream applications Rapid and simple procedureFEATURES AND BENEFITSIsolate DNA from a wide range of samples -Genomic DNAcan be isolated from viable yeast cells, fungal spores or mycelium No phenol:chloroform extractions - DNAis isolated without the use of harmful chemicals such as phenol or chloroform Rapid and simple procedure - Rapid spin-column format allows for the processing of multiple samples in 45 minutes Isolate high yields of purified DNA - Recovered genomic DNA is suitable for downstream applications including PCR, Restriction Fragment Length Polymorphism (RFLP), and Amplified Fragment Length Polymorphism (AFLP)
Catalog No. 50-196-955 Supplier Bioworld Supplier No. 107600981
For the rapid purification of DNA from yeast cells and fungal spores or mycelium No phenol required for isolation Purified DNA fully compatible with sensitive downstream applications Rapid and simple procedureFEATURES AND BENEFITSIsolate DNA from a wide range of samples -Genomic DNAcan be isolated from viable yeast cells, fungal spores or mycelium No phenol:chloroform extractions - DNAis isolated without the use of harmful chemicals such as phenol or chloroform Rapid and simple procedure - Rapid spin-column format allows for the processing of multiple samples in 45 minutes Isolate high yields of purified DNA - Recovered genomic DNA is suitable for downstream applications including PCR, Restriction Fragment Length Polymorphism (RFLP), and Amplified Fragment Length Polymorphism (AFLP)