Learn More
Norgen Biotek Corp Fungi/Yeast Genomic DNA Isolation Kit
Catalog No. NC1040971
Encompass_Preferred
Norgen's Fungi/Yeast Genomic DNA Isolation kit provides rapid spin column purification of genomic DNA from viable yeast cells, fungal spores or mycelium, and Gram-positive and Gram-negative bacteria. Genomic DNA is efficiently extracted from the cells by a combination of heat treatment, detergents and bead tubes (provided). An optional lyticase treatment allows for improved DNA yields with certain fungal and yeast species. Protocol time is typically less than 30 minutes. Recovered genomic DNA is of excellent yield and purity for any downstream application including PCR, qPCR, Restriction Fragment Length Polymorphism (RFLP), Amplified Fragment Length Polymorphism (AFLP), sequencing, SNP analysis and more.
Catalog No. NC1040971 Supplier Norgen Biotek Corp Supplier No. 27300
Norgen's Fungi/Yeast Genomic DNA Isolation kit provides rapid spin column purification of genomic DNA from viable yeast cells, fungal spores or mycelium, and Gram-positive and Gram-negative bacteria. Genomic DNA is efficiently extracted from the cells by a combination of heat treatment, detergents and bead tubes (provided). An optional lyticase treatment allows for improved DNA yields with certain fungal and yeast species. Protocol time is typically less than 30 minutes. Recovered genomic DNA is of excellent yield and purity for any downstream application including PCR, qPCR, Restriction Fragment Length Polymorphism (RFLP), Amplified Fragment Length Polymorphism (AFLP), sequencing, SNP analysis and more.