Learn More
Invivogen FUNGIN 75 MG (10 MG/ML)
SDP
Catalog No. NC9326704
Encompass
Fungin™ kills fungi and yeast by interfering with the cell membrane. The ergosterol, only found in fungi and yeast, is the primary site of action of Pimaricin. This polyene-induced distortion of selective membrane permeability results in a leakage of potassium and magnesium ions and a decrease of macromolecule synthesis. The death of fungi and yeast cells is characterized by the penetration of protons resulting in intracellular acidification. Product concentration: 10 mg/ml Formula: C33H47NO13 Molecular weight: 665.74 Active concentration: from 10 μg/ml to 50 μg/ml Highly stable and water soluble compound Compatible with Pen/Strep solutions No toxicity on treated cells and no deleterious effects on cell metabolism
Catalog No. NC9326704 Supplier Invivogen Supplier No. ANTFN1
Fungin™ kills fungi and yeast by interfering with the cell membrane. The ergosterol, only found in fungi and yeast, is the primary site of action of Pimaricin. This polyene-induced distortion of selective membrane permeability results in a leakage of potassium and magnesium ions and a decrease of macromolecule synthesis. The death of fungi and yeast cells is characterized by the penetration of protons resulting in intracellular acidification. Product concentration: 10 mg/ml Formula: C33H47NO13 Molecular weight: 665.74 Active concentration: from 10 μg/ml to 50 μg/ml Highly stable and water soluble compound Compatible with Pen/Strep solutions No toxicity on treated cells and no deleterious effects on cell metabolism