Learn More
Newcomer Supply Fungus, GMS, Candida sp., Artificial Control Slides, 10/set
SDP
Catalog No. NC9066490
Encompass
Newcomer Supply Fungus, GMS, Candida sp., Artificial Control Slides are for the positive histochemical staining of Candida sp. in tissue sections. Candida albicans purchased from Remel Microbiology Products is used to produce the positive control tissue. Fungus, GMS, Candida sp., Artificial Control Slides contain sections of positive staining rat lung and negative staining human lung.
Catalog No. NC9066490 Supplier Newcomer Supply Supplier No. 4230A
Newcomer Supply Fungus, GMS, Candida sp., Artificial Control Slides are for the positive histochemical staining of Candida sp. in tissue sections. Candida albicans purchased from Remel Microbiology Products is used to produce the positive control tissue. Fungus, GMS, Candida sp., Artificial Control Slides contain sections of positive staining rat lung and negative staining human lung.