Learn More
Newcomer Supply Fungus, GMS, Aspergillus sp., Artificial Control Slides, 10/set
SDP
Catalog No. NC0046687
Encompass
Newcomer Supply Fungus, GMS, Aspergillus sp., Artificial Control Slides are for the positive histochemical staining of Aspergillus sp. in tissue sections. Aspergillus fumigatus purchased from Remel Microbiology Products is used to produce the positive control tissue. Fungus, GMS, Aspergillus sp., Artificial Control Slides contain sections of positive staining rat lung and negative staining human lung.
Catalog No. NC0046687 Supplier Newcomer Supply Supplier No. 4220A
Newcomer Supply Fungus, GMS, Aspergillus sp., Artificial Control Slides are for the positive histochemical staining of Aspergillus sp. in tissue sections. Aspergillus fumigatus purchased from Remel Microbiology Products is used to produce the positive control tissue. Fungus, GMS, Aspergillus sp., Artificial Control Slides contain sections of positive staining rat lung and negative staining human lung.